ContactUsLinkComponent
BioSpectrum 500 System. With BioChemi camera: 12-bit monochrome, digital camera and mount, 2048 x 2048 chip with binning capability (4Mp). Two stage, forced air assisted, peltier cooling to -28C from zero. Camera mount and fixed lens 50mm, f/1.4, clo 1 * 1 items
Catalog # 733-2021
BioSpectrum 500 System. With BioChemi camera: 12-bit monochrome, digital camera and mount, 2048 x 2048 chip with binning capability (4Mp). Two stage, forced air assisted, peltier cooling to -28C from zero. Camera mount and fixed lens 50mm, f/1.4, clo 1 * 1 items
Catalog # 733-2021
Frequently Bought Together