Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Inlet liners

Siltek deactivated glass PSS liner for split operation, 4 mm, 2 mm, 86,2 mm
Catalog # PERCN6502002
Supplier:   PerkinElmer
Inlet liners
Catalog # PERCN6502002
Supplier:  PerkinElmer
Inlet liners

Siltek deactivated glass PSS liner for split operation, 4 mm, 2 mm, 86,2 mm

Price
SFr. 1’100.00
Log in for availability
1 * 1 ST
Product Details & Documents

Liners protect GC columns against column contamination. These glass split and splitless inlet liners promote full vaporisation of samples, minimise mass discrimination, and achieve accurate and reproducible results.

  • Liners packed with wool help to trap non-volatile material
Specifications
  • Ext.Ø:
    4 mm
  • Ø int.:
    2 mm
  • Description:
    Siltek deactivated glass PSS liner for split operation
  • Length:
    86,2 mm
  • Seq:
    06