ContactUsLinkComponent
Rack, flipper 1, colour yellow, material PP, autoclavable, temperature resistant -90°C to +120°C, dimension 197x95x115mm, 8 holes di ameter 30mm, 20 holes diameter 20mm. 1 * 1 items
Catalog # KOEH2-2771
Rack, flipper 1, colour yellow, material PP, autoclavable, temperature resistant -90°C to +120°C, dimension 197x95x115mm, 8 holes di ameter 30mm, 20 holes diameter 20mm. 1 * 1 items
Catalog # KOEH2-2771
Frequently Bought Together