Drum taps, PE
Fast outflow, Corrosive and aggressive chemicals, 3/4" Tri-Sure
MSGA2801
: asecos
These polyethylene taps are used for corrosive and aggressive chemicals.
- Fast outflow
- Tri-sure thread
- Thread:3/4" Tri-Sure
- Description:Fast outflow
- Seq:01
- For:Corrosive and aggressive chemicals