Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Waste Containers

Dispensing bin with swinging lid, Red, 90 L, 400×400×640 mm
Catalog # MSGA8842
Supplier:   asecos
Waste Containers
Catalog # MSGA8842
Supplier:  asecos
Waste Containers
Waste Containers

Dispensing bin with swinging lid, Red, 90 L, 400×400×640 mm

Price
SFr. 324.00
Log in for availability
1 * 1 ST
Product Details & Documents

Made of sheet steel, lacquered.

With swinging lid.

Specifications
  • Capacity:
    90 L
  • Colour:
    Red
  • Description:
    Dispensing bin with swinging lid
  • W×D×H:
    400×400×640 mm