Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Waste Containers

Waste Containers

Dispensing bin with swinging lid, Grey/blue, 90 L, 400×400×640 mm
Catalog # MSGA14873
Supplier:   asecos
Waste Containers
Catalog # MSGA14873
Supplier:  asecos
Waste Containers
Waste Containers

Dispensing bin with swinging lid, Grey/blue, 90 L, 400×400×640 mm

Price
SFr. 324.00
Log in for availability
1 * 1 ST
Product Details & Documents

Made of sheet steel, lacquered.

With swinging lid.

Specifications
  • Capacity:
    90 L
  • Colour:
    Grey/blue
  • Description:
    Dispensing bin with swinging lid
  • W×D×H:
    400×400×640 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.