Back pressure regulator
Biocompatible back pressure regulator, 1 - 20 bar
554-1492
This regulator installs on the detector drain line and prevents bubble formation in the flow cell during outgassing. It has an adjustable back pressure from 0 to 20 bar (15 to 300 psi) without disconnecting from the system. Other sizes available.
- Improves baseline stability
- Low dead volume
- Biocompatible
- Description:Biocompatible back pressure regulator
- Seq:01
- Pressure:1 - 20 bar



