Objectives
A-Planachromatic, Ph2, M27, N.A. of 0,65, WD of 0,45 mm
Catalog # ZEIS4410519910000
Supplier: ZEISS
Jump to:
Product Details & Documents
Specifications
- Magnification:40×
- Description:A-Planachromatic, Ph2, M27, N.A. of 0,65, WD of 0,45 mm
- Seq:12