PIG® Poly Drip Decks
Drip deck, 42 L sump capacity, 680 kg load capacity, White/black, 9,1 kg, 660×660×150 mm
Catalog # 121-0185
Supplier: NEW PIG
Jump to:
Product Details & Documents
These poly drip decks capture drips from containers and drums before they cause slip-and-fall hazards. These decks keep leaky drums and containers from creating puddles and slick spots on the floors.
- Low-profile 15 cm high design is easy to load or unload; pumps and funnels are easy to access
- Molded-in sump catches leaks, drips and spills to help keep your storage areas clean and safe
- Low-density polyethylene (LDPE) construction resists UV rays, rust, corrosion and most chemicals
- Textured grating adds traction and lifts out for easy sump access
- Translucent white sidewalls allow easy visual inspection
Partly made from post-industrial recycled polyethtylene.
Specifications
- Weight:9,1 kg
- Colour:White/black
- Description:Drip deck, 42 L sump capacity, 680 kg load capacity
- Seq:01
- W×D×H:660×660×150 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



