Adapters for Huber circulators
Adapter, M30×1,5 male thread to 1/2" female thread
Catalog # HUBE6394
Supplier: HUBER
Jump to:
Product Details & Documents
Specifications
- Description:Adapter, M30×1,5 male thread to 1/2" female thread
- Seq:24