Orders LinkContactUsLinkComponent
Verification standard kit, MultiDirect
Verification standard kit, Multidirect 0/430/530/560/580/610/660 nm, 1 KIT
Catalog # LOVI215650
Supplier:   Lovibond
Verification standard kit, MultiDirect
Catalog # LOVI215650
Supplier:  Lovibond
CAS Number:  
Verification standard kit, MultiDirect
Verification standard kit, MultiDirect
Verification standard kit, Multidirect 0/430/530/560/580/610/660 nm, 1 KIT
CAS Number:  
Price
SFr. 315.00
Log in for availability
1 * 1 KIT
Product Details & Documents
Verification accuracy of photometers and validity of results.
  • Check the performance of photometers over the full measurement range
  • Results can be checked against tolerances provided on the certificate of conformity
 : Contains zero solution in sealed photometer vial, 6× verification standard solution in sealed photometer vial, light shield cap, certificate of conformity, instruction sheet.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Verification standard kit, Multidirect 0/430/530/560/580/610/660 nm
Specifications