ContactUsLinkComponent
Polyamide/Aluminium/Polyester pouches for gamma sterilisation. Peelable pouches are designed for a quick and easy presentation of sterile items. The three layer laminated film with its 100% aluminium barrier is perfectly suited for sterile items whic 1 * 10.000 items
Catalog # 129-0284
Supplier:  S▄DPACK MEDICA
Polyamide/Aluminium/Polyester pouches for gamma sterilisation. Peelable pouches are designed for a quick and easy presentation of sterile items. The three layer laminated film with its 100% aluminium barrier is perfectly suited for sterile items whic 1 * 10.000 items
Catalog # 129-0284
Supplier:  S▄DPACK MEDICA
Frequently Bought Together