Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Stopcocks, straight bore

2,5 mm, 9 mm
  BIBBSST203
 :   SciLabware
Stopcocks, straight bore
  BIBBSST203
Stopcocks, straight bore
Stopcocks, straight bore

2,5 mm, 9 mm

 
SFr. 549.00
   
1 * 5 ST
 

PYREX®, borosilicate glass with PTFE key.

  • Barrels are highly polished and the keys are self lubricating, avoiding the need for stopcock grease
  • Keys are fully interchangeable and supplied complete with plastic screw thread retaining device
  • Made to a taper of 1:10 giving a combination of performance and ease of operation
 
  • Hole Ø:
    2,5 mm
  • Seq:
    06
  • For tube I-Ø:
    9 mm
Accessories

 

  •  
    Description
    Hole Ø
     
     
  • BIBBSST102K
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    1,5 mm
       
    1 * 5 ST - SFr. 124.00

 

  •  
    Description
    Hole Ø
     
     
  • BIBBSST103K
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    2,5 mm
       
    1 * 5 ST - SFr. 145.00

 

  •  
    Description
    Hole Ø
     
     
  • BIBBSST104K
    Spare PTFE keys for stopcocks, PYREX®, straight bore
    4,0 mm
       
    1 * 5 ST - SFr. 206.00

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.