Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Volumetric flasks, class B, blue markings

Volumetric flasks, class B, blue markings

500 ml, 19/26, 260 mm, 0,50 ± ml
Catalog # BIBBFRD036P
Supplier:   SciLabware
Volumetric flasks, class B, blue markings
Catalog # BIBBFRD036P
Supplier:  SciLabware
Volumetric flasks, class B, blue markings
Volumetric flasks, class B, blue markings

500 ml, 19/26, 260 mm, 0,50 ± ml

Price
SFr. 104.00
Log in for availability
1 * 2 ST
Product Details & Documents

MBL®, borosilicate glass, with PE stopper.

  • Marks and inscriptions in high contrast blue enamel
  • Excellent temperature resistance

Certifications: Complies with ISO 1042 and DIN 12664

Specifications
  • NS:
    19/26
  • Capacity:
    500 ml
  • Height:
    260 mm
  • Seq:
    09
  • Tolerance:
    0,50 ± ml
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.