Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Spare keys for stopcocks, Pyrex®

Spare keys for stopcocks, Pyrex®

Replacement keys, for 12,5 mm Ø, 1,5 mm, SSD series stopcocks
Catalog # BIBBSSD102K
Supplier:   SciLabware
Spare keys for stopcocks, Pyrex®
Catalog # BIBBSSD102K
Supplier:  SciLabware
Spare keys for stopcocks, Pyrex®
Spare keys for stopcocks, Pyrex®

Replacement keys, for 12,5 mm Ø, 1,5 mm, SSD series stopcocks

Price
39.00 SFr.
Log in for availability
1 * 1 ST
Product Details & Documents

Spare keys for stopckocks, made of borosilicate glass.

  • Straight bore
  • Interchangeable

Delivery information: Supplied complete with key retaining device.

Specifications
  • For:
    SSD series stopcocks
  • Hole Ø:
    1,5 mm
  • Description:
    Replacement keys, for 12,5 mm Ø
  • Seq:
    01
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.