Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Brushes for test tubes

Brushes for test tubes

Test tube brush, with fallow bristles, Hog bristle, 95 mm, 21 mm, Wire, 230 mm
Catalog # REIT12230301
Brushes for test tubes
Catalog # REIT12230301
Brushes for test tubes

Test tube brush, with fallow bristles, Hog bristle, 95 mm, 21 mm, Wire, 230 mm

Price
SFr. 12.50
Log in for availability
1 * 1 ST
Product Details & Documents

Brushes with natural bristles and zinc plated wire handle.

Specifications
  • Bristle material:
    Hog bristle
  • Brush head Ø:
    21 mm
  • Handle material:
    Wire
  • Description:
    Test tube brush, with fallow bristles
  • Seq:
    19
  • Total length:
    230 mm
  • Brush head length:
    95 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.