ContactUsLinkComponent
Chemical Process Indicator Label for monitoring Radiation sterilization processes. Labels are 4.5 x 3 with Class 1 ISO 11140-1: 2005 compliant indicating strip. Labels are printable by thermal transfer and provide visual assurance that a product 1 * 1 Roll
Catalog # NAMSSP-R07
Supplier:  CROSSTEX
Chemical Process Indicator Label for monitoring Radiation sterilization processes. Labels are 4.5 x 3 with Class 1 ISO 11140-1: 2005 compliant indicating strip. Labels are printable by thermal transfer and provide visual assurance that a product 1 * 1 Roll
Catalog # NAMSSP-R07
Supplier:  CROSSTEX
Frequently Bought Together