Orders LinkContactUsLinkComponent
Tripod stands
120×260 mm
  451-0161
Tripod stands
  451-0161
Tripod stands
Tripod stands
120×260 mm
50.70 SFr.
1 * 1 ST
Tripod stands made of stainless steel or zinc plated steel.
Please note: Zinc plating of tripod stands serves as temporary corrosion protection. May peel off after direct contact with open burner flame.
  • Int.Ø×H:
    120×260 mm
  • Seq:
    04