Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Formamide ≥99%

Formamide ≥99%

Glass bottle for solids, 10.000 g
Catalog # A11076.0C
Health
Formamide ≥99%
Catalog # A11076.0C
CAS Number:  75-12-7
Formamide ≥99%
Formamide ≥99%

Glass bottle for solids, 10.000 g

CAS Number:  75-12-7
Price
SFr. 808.00
Log in for availability
1 * 10.000 g
Product Details & Documents

Purity: 99% CAS No.: 75-12-7 Molecular Formula: CH3NO Formula Weight: 45.04 Flash Point: 150°(302°F) Refractive Index: 1.4475 Odour: Ammonia-like Melting Point: 2° Boiling Point: 210° Beilstein Reference: 505995 Merck Reference: 14,4237 MDL No.: MFCD00007941

Synonyms:  Carbamaldehyde
  • CAS Number:75-12-7
  • Formula:HCONH₂
  • MDL Number:MFCD00007941
  • Molecular Weight:45.04 g/mol
  • Merck Index:14,4237
  • Boiling Pt:210 °C
  • Melting Pt:2 °C
  • Density:1.134
  • Flash Pt:150 °C (302 °F)
  • Storage Temperature:Ambient
  • EINECS:200-842-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10.000 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.