Stopcocks, straight bore
4 mm, 10 mm
201-0569
PYREX®, borosilicate glass with PTFE key.
- Barrels are highly polished and the keys are self lubricating, avoiding the need for stopcock grease
- Keys are fully interchangeable and supplied complete with plastic screw thread retaining device
- Made to a taper of 1:10 giving a combination of performance and ease of operation
- Hole Ø:4 mm
- Seq:03
- For tube I-Ø:10 mm
Accessories
- DescriptionHole Ø
- DescriptionHole Ø
- DescriptionHole Ø
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



