Phosphate Buffer, pH 6.6 at 25 °C
Phosphate buffer, 6,6, −, 12
P8165-12VL
Phosphate buffer's buffering capacity is pH sensitive and is preferred to be used at physiological pH (7,2 to 7,4).
- Each vial will prepare 3,8 L of buffer, 34 mM
- Used in hematology and histology
For use with Wright Stain for Geometric Data HEMASTAIN®. Phosphate buffer serves as an important fixative for light and electron microscopy. It is the most commonly used component of irrigation solution during surgeries, to preserve living tissues. It is also used in immunochemical staining procedures.
- Pack type:−
- Pk:12
- Description:Phosphate buffer
- pH value:6,6
- Cat. No.:P8165-12VL
- M-Clarity quality level:MQ200
- Storage temperature:RT
- Appearance:White powder



