ContactUsLinkComponent
Microloops© D Sterile disposable plastic 5?l inoculating loops for precise volumetric sampling and transfer of material for accurate analysis. Packed in tamper evident resealable polythene sleeves, 20 loops per sleeve. Colour: Purple 1 * 1.000 items
Catalog # MEDWMW705/20
Microloops© D Sterile disposable plastic 5?l inoculating loops for precise volumetric sampling and transfer of material for accurate analysis. Packed in tamper evident resealable polythene sleeves, 20 loops per sleeve. Colour: Purple 1 * 1.000 items
Catalog # MEDWMW705/20
Frequently Bought Together