Pycnometer with thermometer, adjusted
50 ml, 10/19
Catalog # HECH42572032
Supplier: GLASWARENFABRIK KARL HECHT
Jump to:
Product Details & Documents
Pycnometer, made of borosilicate glass 3.3, Gay-Lussac type.
- With ground-in capillary stopper
- Very good chemical resistance
- Very good temperature resistance
It includes pycnometer bottle and ground-in thermometer joint size 10/19 with blue filling.
Certifications: DIN ISO 3507
Documents
Specifications
- NS:10/19
- Volume (nom.):50 ml