ContactUsLinkComponent
Filter paper, quantitative, diameter 240mm, grade 391, format folded filters, thickness 0.15mm, particle retention 2-3µm, precipitat ion very fine crystalline, colour coded dot blue, made of refined pulp and linters with over 98% alpha-cellulose cont 1 * 100 items
Catalog # SARTFT-4-104-240
Supplier:  Sartorius
Filter paper, quantitative, diameter 240mm, grade 391, format folded filters, thickness 0.15mm, particle retention 2-3µm, precipitat ion very fine crystalline, colour coded dot blue, made of refined pulp and linters with over 98% alpha-cellulose cont 1 * 100 items
Catalog # SARTFT-4-104-240
Supplier:  Sartorius
Frequently Bought Together