Qualitative filter papers, technical, smooth, grade 3 m/N
Product Details & Documents
These smooth filter papers are made of refined pulp and linters with more than 95% alpha-cellulose content. The ash content of these filter papers is between 0,1 and 0,15%. They are used for routine analyses like clarification, determination of substances, but also as discs with a centre hole for technical applications. Grades with a wet burst resistance >30 kPa are referred to as wet-strengthened and are therefore suitable for pressure or vacuum filtration.
Medium fast rate of filtration, thin, smooth, increased wet strength. Used for acids; general vacuum or pressure filtration applications and less critical analyses; also used to assay the sucrose content in cane sugar solutions with a relatively low viscosity.
Specifications
- Grade:3 m/N
- Seq:02
- Size:110 mm
- Thickness:0,14 mm
- Cat. No.:SARTFT-3-305-110
- Wet burst strength:30 kPa
- Environmental certification:Renewable Energy Certificate
- Filtration speed:30 s
- Particle retention:7 - 10 µm
- Environmentally Preferable:Low Manufacturing Impact
- Weight:65 g/m²
Les recommandations seront personnalisées selon vos préférences d’achat uniquement si vous avez activé "Améliorer mon expérience d’achat" dans la page "Informations personnelles".
Sinon, vous recevrez des recommandations génériques.



