ContactUsLinkComponent
Filter, disc diameter 240mm, material glass microfiber, grade MGB, binder-free, temperature resistant up to 500°C or 550 °C, highly pure and extremely white. 1 * 25 items
Catalog # SARTFT-3-1102-240
Supplier:  Sartorius
Filter, disc diameter 240mm, material glass microfiber, grade MGB, binder-free, temperature resistant up to 500°C or 550 °C, highly pure and extremely white. 1 * 25 items
Catalog # SARTFT-3-1102-240
Supplier:  Sartorius
Frequently Bought Together