ContactUsLinkComponent
Filter, disc diameter 55mm, material glass microfiber, grade MGB, binder-free, temperature resistant up to 500°C or 550 °C, highly p ure and extremely white. 1 * 50 items
Catalog # SARTFT-3-1102-055
Supplier:  Sartorius
Filter, disc diameter 55mm, material glass microfiber, grade MGB, binder-free, temperature resistant up to 500°C or 550 °C, highly p ure and extremely white. 1 * 50 items
Catalog # SARTFT-3-1102-055
Supplier:  Sartorius
Frequently Bought Together