ContactUsLinkComponent
Seal, cap size 20mm, crimp, material aluminum, cap colour silver, septum gray PTFE/black butyl rubber, pharmafix, operating temperat ure -40 to +125?C, septum provides improved sealing especially for square edge vials
Catalog # CRMAC4020-36A
Supplier:  CHROMACOL
Seal, cap size 20mm, crimp, material aluminum, cap colour silver, septum gray PTFE/black butyl rubber, pharmafix, operating temperat ure -40 to +125?C, septum provides improved sealing especially for square edge vials
Catalog # CRMAC4020-36A
Supplier:  CHROMACOL