ContactUsLinkComponent
Insert, volume 6mm, material silanized glass, mandrel tip, conical, usable volume 500µl, residual volume <8µl, flanged lip, improves centering of lip, silanized glass reduces non-specific binding
Catalog # CRMAC4015-S641
Supplier:  CHROMACOL
Insert, volume 6mm, material silanized glass, mandrel tip, conical, usable volume 500µl, residual volume <8µl, flanged lip, improves centering of lip, silanized glass reduces non-specific binding
Catalog # CRMAC4015-S641
Supplier:  CHROMACOL