ContactUsLinkComponent
Seal, cap size 11mm, crimp, material aluminum, cap colour green, septum clear PTFE/red rubber, most commonly used GC septum
Catalog # CRMAC4011-98G
Supplier:  CHROMACOL
Seal, cap size 11mm, crimp, material aluminum, cap colour green, septum clear PTFE/red rubber, most commonly used GC septum
Catalog # CRMAC4011-98G
Supplier:  CHROMACOL