ContactUsLinkComponent
Septum, 10mm, target, white, virgin, PTFE, designed to used with 10-425 threaded caps
Catalog # CRMAC4010-10
Supplier:  CHROMACOL
Septum, 10mm, target, white, virgin, PTFE, designed to used with 10-425 threaded caps
Catalog # CRMAC4010-10
Supplier:  CHROMACOL