Orders LinkContactUsLinkComponent
Goggles, Flex Seal™
Goggles, Flex Seal™
Anti-UV, anti-scratch, anti-fog, Clear PC, Blue/grey
Catalog # 111-3606
Supplier:   HONEYWELL SAFETY
Goggles, Flex Seal™
Catalog # 111-3606
Supplier:  HONEYWELL SAFETY
Goggles, Flex Seal™
Goggles, Flex Seal™
Anti-UV, anti-scratch, anti-fog, Clear PC, Blue/grey
Price
SFr. 41.80
Log in for availability
1 * 1 ST
Product Details & Documents
Goggles with a flexible and sturdy frame feature a wraparound lens for a panoramic view.
  • Soft and flexible silicone frame for excellent comfort and ensures a snug, slip-free fit
  • Remains soft and supple even in extremely hot or cold environments
  • Pivoting, removable and adjustable headband
  • Indirect ventilation prevents fogging
  • Neoprene strap (111-0132) or elastic strap (111-3606)
Designed to be compatible with prescription spectacles.
Weight: 110 g.
 : EN 166, EN 169, EN 171, 1FT, C 3, 4, 9.
Specifications
  • Type:
    Anti-UV, anti-scratch, anti-fog
  • Colour:
    Blue/grey
  • Lenses:
    Clear PC