Calibration weights, OIML M3
5000 g, With certificate
Catalog # OHAU11117245
Supplier: OHAUS
Jump to:
Product Details & Documents
Test weights made from stainless steel with adjusting cavity.
Delivery information: Supplied in a plastic box.
Specifications
- Weight:5000 g
- Seq:12
- Type:With certificate