Calibration weights, OIML M3
2 g, With certificate
Catalog # OHAU11117235
Supplier: OHAUS
Jump to:
Product Details & Documents
Test weights made from stainless steel with adjusting cavity.
Delivery information: Supplied in a plastic box.
Specifications
- Weight:2 g
- Seq:02
- Type:With certificate