Calibration weights, OIML M3
200 g, Without certificate
Catalog # OHAU11117213
Supplier: OHAUS
Jump to:
Product Details & Documents
Test weights made from stainless steel with adjusting cavity.
: Supplied in a plastic box.
Specifications
- Weight:200 g
- Seq:21
- Type:Without certificate