L(+)-Lithium lactate ≥97%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
L(+)-Lithium lactate ≥97%
Catalog # A11818.0B
Supplier: Thermo Fisher Scientific
CAS Number: 27848-80-2
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 27848-80-2 Molecular Formula: C3H5LiO3 Formula Weight: 96.01 Melting Point: 253-255° Beilstein Reference: 3568332
Synonyms: Lithium (S)-2-hydroxypropanoate, (S)-(+)-Lactic acid lithium salt, (S)-2-Hydroxypropionic acid Lithium salt, Lithium (S)-2-Hydroxypropionate, L(+)-Lactic acid lithium salt, (S)-(+)-2-Hydroxypropanoic acid lithium salt, (S)-(+)-2-Hydroxypropionic acid lithium salt, Lithium L-Lactate, L-Lactic acid Lithium salt
- CAS Number:27848-80-2
- Formula:C₃H₅LiO₃
- MDL Number:MFCD00065512
- Molecular Weight:96.01 g/mol
- Melting Pt:253…255 °C
- Storage Temperature:Ambient
- EINECS:248-692-5
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



