Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Micro extraction by packed sorbent (MEPS™)

60 mm scale, 6,7 mm OD barrel, 250 µl, HTA 300APlus, Thermo Scientific & Varian 8400
  554-1459
Micro extraction by packed sorbent (MEPS™)
  554-1459
Micro extraction by packed sorbent (MEPS™)
Micro extraction by packed sorbent (MEPS™)

60 mm scale, 6,7 mm OD barrel, 250 µl, HTA 300APlus, Thermo Scientific & Varian 8400

 
SFr. 227.00
   
1 * 1 ST
 

Micro Extraction by Packed Sorbent (MEPS™) is a new development in the field of sample preparation by SPE (solid phase extraction). The MEPS™ product consists of two parts, the MEPS™ syringe and the MEPS™ Barrel Insert and Needle assembly (BIN). The MEPS™ approach to sample preparation is suitable for reversed phases, normal phases, mixed mode and ion exchange chemistries. Because MEPS™ BINs can be used for sample volumes as small as 10 µl it makes them particularly well suited to on-line use with LC-MS analysis of volume limited samples.

  • The BIN contains the SPE phase that is built into the syringe needle
  • The BIN is used with a 100 or 250 µl gas tight MEPS™ syringe that allows fluid handling at normal SPE pressures
  • Sample preparation for complex biological samples is readily adapted to MEPS™ and reduces the volume of sample and reagents required for extraction

Base material is silica with mean particle size of 45 µm and pore size of 60 Å.

 
  • For:
    HTA 300APlus, Thermo Scientific & Varian 8400
  • Volume:
    250 µl
  • Description:
    60 mm scale, 6,7 mm OD barrel
  • Seq:
    03
Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..