Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Coloured slide boxes

Slide box with snap lock, Violet, PS, 25, 90×90×32 mm
  GLWAK25V
 :   GLW
Coloured slide boxes
  GLWAK25V
 :  GLW
Coloured slide boxes
Coloured slide boxes

Slide box with snap lock, Violet, PS, 25, 90×90×32 mm

 
SFr. 7.80
   
1 * 1 ST
 

Slide boxes with 25 compartments and snap lock provide a secure hold for the slides and coverslips.

  • Stackable for use as a minimum space as possible
  • Can be used in a freezer down to –80 °C
  • Numbers in the bottom of the box and hinged lid

Designed for storing, transporting, and freezing the samples.

 : Dividers must be ordered separately.

 
  • No. of slides:
    25
  • Colour:
    Violet
  • Description:
    Slide box with snap lock
  • Material:
    PS
  • Seq:
    07
  • W×D×H:
    90×90×32 mm
Accessories

 

  •  
    Description
    Colour
    Material
     
     
  • GLWAZK25
    Divider for slides and coverslips of special sizes
    White
    PS
       
    1 * 1 ST - SFr. 3.00

 

  •  
    Description
    Colour
    Material
     
     
  • GLWAZK20
    Divider for slides and coverslips of special sizes
    Grey
    PS
       
    1 * 1 ST - SFr. 3.10

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.