Staining tiles
24, Ø×H: 24×3 mm, Transparent, 130×200×6 mm
Catalog # MARI1406703
Supplier: MARIENFELD
Jump to:
Product Details & Documents
- Bevelled edges
- Ground and polished cavities
- Clipped corners: Except the one with twelve wells
Specifications
- No. of wells:24
- Colour:Transparent
- W×D×H:130×200×6 mm
- Well size:Ø×H: 24×3 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



