Order Entry
Switzerland
Orders LinkContactUsLinkComponent
HEPES Buffer Solution 1 M, HyClone™

HEPES Buffer Solution 1 M, HyClone™

HyClone HEPES 1M Solution, Glass bottle, 100 ml
Catalog # HYCLSH30237.01
HEPES Buffer Solution 1 M, HyClone™
Catalog # HYCLSH30237.01
CAS Number:  
HEPES Buffer Solution 1 M, HyClone™
HEPES Buffer Solution 1 M, HyClone™

HyClone HEPES 1M Solution, Glass bottle, 100 ml

CAS Number:  
Price
SFr. 28.50
Log in for availability
1 * 100 mL
Product Details & Documents

Maintain physiological pH in cell culture, despite changes in carbon dioxide concentration (produced by cellular respiration) with Cytiva HyClone HEPES solution.

  • 0,1 µm sterile filtered
  • pH: 7,2 - 7,5
  • Storage: 2 to 8° C

Certifications: Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities

Documents

Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    100 ml
  • Description:
    HyClone HEPES 1M Solution

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.