Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

Tripod stands

Retort stand base, tripod, 0,68 kg, 80 mm
Catalog # 241-0377
Supplier:   USBECK Laborgeräte
Tripod stands
Catalog # 241-0377
Tripod stands
Tripod stands

Retort stand base, tripod, 0,68 kg, 80 mm

Price
SFr. 32.40
Log in for availability
1 * 1 ST
Product Details & Documents

Steel, powder coated.

  • With M10 threaded hole
  • No support rod
  • Non-slip, rubber feet ensure firm footing
Specifications
  • Description:
    Retort stand base, tripod
  • Arm length:
    80 mm
  • Weight:
    0,68 kg