Orders LinkContactUsLinkComponent
Microarray slides, HTA™
Microarray slides, HTA™
3-D aldehyde microarray slides, 1
Catalog # 731-3010
Supplier:   Greiner Bio-One
Microarray slides, HTA™
Catalog # 731-3010
Supplier:  Greiner Bio-One
Microarray slides, HTA™
Microarray slides, HTA™
3-D aldehyde microarray slides, 1
Price
SFr. 377.00
Log in for availability
1 * 5 ST
Product Details & Documents
The HTA™ 1 can be used for the processing of one sample. The HTA™ 12 are partitioned into 12 flat compartments, each with a printable surface of 6×6 mm and a low well rim of 0,5 mm. Twelve samples can be processed simultaneously.
  • Multifunctional 3-D amino, 3-D aldehyde or 3-D epoxy surface functionality
  • Low well rim of only 0,5 mm, low auto-fluorescence
  • Plastic plates for high-throughput microarrays
Combine the advantages of the universal microplate platform with microarrays for diagnostic applications due to identical well geometry compared with a standard 96-well microplate.
Specifications
  • No. of wells:
    1
  • Description:
    3-D aldehyde microarray slides