Orders LinkContactUsLinkComponent
Microarray plates, HTA™ Plate
Microarray plates, HTA™ Plate
HTA Plate, 3-D Amino
Catalog # 731-3018
Supplier:   Greiner Bio-One
Microarray plates, HTA™ Plate
Catalog # 731-3018
Supplier:  Greiner Bio-One
Microarray plates, HTA™ Plate
Microarray plates, HTA™ Plate
HTA Plate, 3-D Amino
Price
1 890.00 SFr.
Log in for availability
1 * 5 ST
Product Details & Documents
Plastic plates for high-throughput microarrays. With 96 shallow wells and a low well rim, the HTA™ Plate has been optimised for the simultaneous analysis of multiple samples.
  • Available with multifunctional, 3-D Amino, 3-D Aldehyde or 3-D Epoxy surface functionality
  • Partition of the plate into four individual strips containing 24 wells each gives flexibility, permitting processing of a variable number of samples each day
  • Plate dimensions are compatible with automated systems
  • Low well rim of only 0,3 mm
  • Low autofluorescence
Specifications
  • Description:
    HTA Plate, 3-D Amino