Order Entry
Switzerland
Orders LinkContactUsLinkComponent
qScript™ One-Step qRT-PCR kits

qScript™ One-Step qRT-PCR kits

qScript™ One-Step qRT PCR Kit, without passive reference dye, 200 reactions, Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480, 200 Assays
Catalog # 733-1257
Supplier:   Quantabio
qScript™ One-Step qRT-PCR kits
Catalog # 733-1257
Supplier:  Quantabio
CAS Number:  
qScript™ One-Step qRT-PCR kits
qScript™ One-Step qRT-PCR kits

qScript™ One-Step qRT PCR Kit, without passive reference dye, 200 reactions, Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480, 200 Assays

CAS Number:  
Price
SFr. 715.00
Log in for availability
Product Details & Documents

Synthesis of cDNA and PCR amplification are carried out in the same tube without opening between procedures. These kits are optimised to deliver maximum RT-PCR efficiency and specificity, enabling unbiased co-amplification of low copy transcripts in the presence of higher copy reference genes. As a result the qScript™ One-Step qRT-PCR Kits offer improved sensitivity over other commercially available qPCR reagents. The qScript™ One-Step Master Mix maximises the activities of both reverse transcriptase and PCR enzymes, while minimising primer-dimer formation. High specificity is assured with AccuStart™ Taq DNA Polymerase, which is bound by monoclonal antibodies for superior hot-start. Upon heating to 95 °C, the antibodies denature irreversibly releasing fully active and unmodified Taq DNA polymerase. qScript™ One-Step qRT-PCR Kits are available for all Real-Time PCR instrument platforms, including those requiring normalisation with ROX reference dye.

  • Broad dynamic range ensures reliable data
  • Highly sensitive for low copy RNA quantification
  • Unbiased duplex amplification for more accurate results
  • Simple, fast protocol with less variability

Documents

Specifications

Specifications

  • Cycler compatibility:
    Agilent AriaMx, BioRad CFX/iCycler iQ™/MyiQ™/iQ™5, QIAGEN Rotor-Gene Q, Quantabio Q, Roche LightCycler 480
  • Pk:
    200 Assays
  • Description:
    qScript™ One-Step qRT PCR Kit, without passive reference dye, 200 reactions
  • Cat. No.:
    733-1257
  • One step:
    Yes
  • Application:
    RT-qPCR probe

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.