Order Entry
Switzerland
Orders LinkContactUsLinkComponent
(S)-(-)-Limonene, Supelco®

(S)-(-)-Limonene, Supelco®

Glass bottle, 1 mL
Catalog # 62128-1ML
Flammable
Irritant
Environment
Supplier:   Merck
(S)-(-)-Limonene, Supelco®
Catalog # 62128-1ML
Supplier:  Merck
CAS Number:  5989-54-8
(S)-(-)-Limonene, Supelco®
(S)-(-)-Limonene, Supelco®

Glass bottle, 1 mL

CAS Number:  5989-54-8
Price
SFr. 66.90
Log in for availability
1 * 1 mL
Product Details & Documents
Synonyms:  (-)-Limonene, (S)-1-Methyl-4-(prop-1-en-2-yl)cyclohex-1-ene, L(-)-Limonene, (S)-(-)-p-Mentha-1,8-diene, (S)-(-)-Dipentene, (S)-(-)-4-Isopropenyl-1-methyl-1-cyclohexene, (S)-(-)-1-Methyl-4-(1-methylvinyl)cyclohexene
  • CAS Number:5989-54-8
  • Formula:C₁₀H₁₆
  • MDL Number:MFCD00001558
  • Molecular Weight:136.24 g/mol
  • Merck Index:13,05515
  • Boiling Pt:175 °C (1013 hPa)
  • Melting Pt:–74 °C
  • Density:0.84 g/cm³ (20 °C)
  • Flash Pt:43 °C
  • Storage Temperature:Ambient
  • UN:2052
  • ADR:3,III
  •  :M-Clarity™ quality level = MQ100
  • EINECS:227-815-6
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 mL

Specifications

Consigli

I suggerimenti saranno personalizzati in base alle tue preferenze di acquisto solo se hai attivato "Migliora la mia esperienza di acquisto" nella pagina "Informazioni personali".

Altrimenti riceverai suggerimenti generici..