Orders LinkContactUsLinkComponent
Rack overlays for sample racks
Rack overlays for sample racks
Rack overlay kit for 20 mm OD tube rack (allows 40, 18 mm O.D. tubes to be used in the rack)
Catalog # VARI6610026000
Supplier:   VARIAN
Rack overlays for sample racks
Catalog # VARI6610026000
Supplier:  VARIAN
Rack overlays for sample racks
Rack overlays for sample racks
Rack overlay kit for 20 mm OD tube rack (allows 40, 18 mm O.D. tubes to be used in the rack)
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Description:
    Rack overlay kit for 20 mm OD tube rack (allows 40, 18 mm O.D. tubes to be used in the rack)
  • Seq:
    02