ContactUsLinkComponent
tranfer rnas (trnas) are used as carriers or co-precipitants for helping in recovery of nucleic acids when alcohol precipitation met hod is used. carrier or co-precipitants are inert substances that enhance recovery of nucleic acid when alcohol is us 1 * 0,5 mL
Catalog # GENO786-058
Supplier:  G-Biosciences
tranfer rnas (trnas) are used as carriers or co-precipitants for helping in recovery of nucleic acids when alcohol precipitation met hod is used. carrier or co-precipitants are inert substances that enhance recovery of nucleic acid when alcohol is us 1 * 0,5 mL
Catalog # GENO786-058
Supplier:  G-Biosciences