Order Entry
Switzerland
Orders LinkContactUsLinkComponent
 

J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate

DILUT-IT™ concentrate, 5,00, ±0,02, Citrate, 20 °C, 1
Catalog # 4853.
Supplier:   Avantor
J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate
Catalog # 4853.
Supplier:  Avantor
CAS Number:  
J.T.Baker® DILUT-IT™, Buffer Solutions, Concentrate

DILUT-IT™ concentrate, 5,00, ±0,02, Citrate, 20 °C, 1

CAS Number:  
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents

Volumetric concentrate, for dilution to 500 ml.

  • Traceable to NIST
Specifications

Specifications

  • pH buffer type:
    Citrate
  • Pk:
    1
  • Reference temperature:
    20 °C
  • Description:
    DILUT-IT™ concentrate
  • pH accuracy:
    ±0,02
  • pH value:
    5,00

Specifications