Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Coagulation tubes, BD Vacutainer®

Coagulation tubes, BD Vacutainer®

Blood Collection tube, BD Vacutainer®, Plastic, BD Hemogard™, Sodium citrate (0,109 M), 75 mm, 13 mm, Light blue, 2,7 ml
Catalog # BDAM363048
Supplier:   BD Medical
Coagulation tubes, BD Vacutainer®
Catalog # BDAM363048
Supplier:  BD Medical
Coagulation tubes, BD Vacutainer®

Blood Collection tube, BD Vacutainer®, Plastic, BD Hemogard™, Sodium citrate (0,109 M), 75 mm, 13 mm, Light blue, 2,7 ml

Price
649.00 SFr.
Log in for availability
1 * 1.000 ST
Product Details & Documents

PET/PP or glass.

  • With buffered citrate
  • With paper lable
  • Hemogard™ closure

Citrate tubes

Specifications
  • Closure type:
    BD Hemogard™
  • Ø:
    13 mm
  • Volume:
    2,7 ml
  • Description:
    Blood Collection tube, BD Vacutainer®
  • Length:
    75 mm
  • Material:
    Plastic
  • Cap colour:
    Light blue
  • Additive:
    Sodium citrate (0,109 M)