Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Drying tubes, U-shaped

Drying tubes, U-shaped

19/26 NS, 250 mm, 18 mm
Catalog # WITG2.114.250
Supplier:   witeg Labortechnik
Drying tubes, U-shaped
Catalog # WITG2.114.250
Supplier:  witeg Labortechnik
Drying tubes, U-shaped
Drying tubes, U-shaped

19/26 NS, 250 mm, 18 mm

Price
SFr. 157.00
Log in for availability
1 * 1 ST
Product Details & Documents

Borosilicate glass, clear. Replaceable NS stopcocks. Lateral tube connectors.

  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Specifications
  • Ø int.:
    18 mm
  • Socket size:
    19/26 NS
  • Length:
    250 mm